The Tychem 4000 Protective Coverall delivers effective protection against a range of chemical environments. It utilizes Saranex 23-P film laminated to Tyvek fabric, creating a rugged and durable material that resists abrasion. This coverall provides at least 30 minutes of protection against >124 chemical challenges, making it suitable for applications like chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
- Features a collar, open wrists, and open ankles for comfort and mobility.
- Storm flap with adhesive closure covers zipper to prevent intrusion.
- Bound seams provide strong, stress-resistant protection.
- Constructed from Saranex 23-P film laminated to Tyvek fabric for durability.
- Manufactured without components containing natural rubber latex.
- Packaged 12 per case.