Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Irradiated Disposable Bag for Mop Head Removal Tool
BIOPHARMAGREENER_CHOICE

Catalog No. 19168575 Shop All Contec Inc. Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-168-575 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-168-575 Supplier Contec™ Supplier No. BNBG0002
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Contec Compact Stainless Steel Bucket System (2940P3S).

  • Cleanroom bag
  • Constructed of 4 mil LDPE
  • Each waste disposal bag will hold approximately 10 VertiKlean™ MAX Sealed Edge and 6 VertiKlean MAX used mops

Specifications

Product Type Disposable Cleanroom Bag
Color Clear
Dimensions 26 x 18 in.
Disposable Single-use
Includes 80 bags/ Case
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.