Learn More
Cardinal Health™ Secure-Gard™ Earloop Procedure Mask
Catalog No. 191201243 Shop All Cardinal Health Products
Change view
Click to view available options
With earloops; Polypropylene; Blue
Catalog No.
19-120-1243 With earloops; Polypropylene; Blue
1 options
This item is not returnable. View return policy
Catalog No. 19-120-1243 Supplier Cardinal Health™ Supplier No. AT7511
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Earloop fastened procedure mask is ideal for quick donning.
  • Earloop design for fast donning
  • Polypropylene outer and inner facing more likely to repel small amounts of fluid such as respiratory droplets
  • Delivers same level of protection if worn correctly or inside out
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color Blue
Fastening Type Earloops
Material Polypropylene
Size Standard
Style Pleated
Latex Free Yes
Model AT7511