Learn More
MSC Dupont Shoe Cover, Non Chemical-Resistant, Polypropylene, Elastic Closure, 200/Pack
Catalog No. 5000793476
Encompass
This shoe cover is designed for general purpose use, offering protection in environments where chemical resistance is not required. Made from polypropylene, it features an elastic closure for a secure fit.
  • Non-chemical-resistant
  • Made from polypropylene material
  • Elastic closure for a secure fit
  • Serged seams for durability
  • Meets ASTM D1894 and ASTM D3776 standards
  • Measures 5.50 in high
Catalog No. 50-007-93476 Supplier MSC Supplier No. 62862743
This shoe cover is designed for general purpose use, offering protection in environments where chemical resistance is not required. Made from polypropylene, it features an elastic closure for a secure fit.
  • Non-chemical-resistant
  • Made from polypropylene material
  • Elastic closure for a secure fit
  • Serged seams for durability
  • Meets ASTM D1894 and ASTM D3776 standards
  • Measures 5.50 in high