Learn More
DuPont™ Sierra™ Controlled Environment Masks
BIOPHARMA
Catalog No. 19171025 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size: 9 in. (22.86cm)
Catalog No.
19-171-025 Size: 9 in. (22.86cm)
1 options
This item is not returnable. View return policy
Catalog No. 19-171-025 Supplier DuPont™ Supplier No. ML7360WH0002500S
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Cleanroom masks provide a balance of particle filtration efficiency and breathability.
  • Pleated
  • Rayon outer facing
  • Metal nose piece
  • Sterile
  • White
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Ties
Sterility Sterile
Style Pleated
Model ML7360WH0002500S