The DuPont™ Tychem® 2000 Sleeved Apron provides lightweight, durable frontal protection against a variety of industrial chemical threats. This yellow, long-sleeved apron is suitable for light liquid splash protection in chemical processing, petroleum refining, pharmaceutical manufacturing, and food processing environments.
- Lightweight and durable Tychem® 2000 fabric offers at least 30 minutes of protection against over 40 chemical challenges.
- Calf-length (52 in) design with attached sleeves ensures enhanced frontal protection.
- Bound seams are tightly sewn and covered to reinforce and reduce particle penetration.
- Open back design for wearer comfort and waist ties for a secure fit.
- Features elastic wrists and a bound yoke neck opening without snaps.
- Manufactured without natural rubber latex.
- One size fits most with a fabric thickness of 10 mils.