Learn More
MSC DuPont Sleeved Apron, Tychem 5000, Taped Seams, 6/Case
Catalog No. 5000673861
Encompass
The DuPont™ Tychem® 5000 Sleeved Apron offers heavy-duty frontal protection against a wide variety of chemical hazards. This knee-length apron features attached sleeves, an open back for comfort, and a tan color for discretionary use.
  • Knee-length apron with attached sleeves
  • Neck opening with snap closure and waist ties
  • Elastic wrists
  • Taped seams
  • Open back for wearer comfort
  • Constructed from multi-layer film barrier laminated to 2.0 oz/yd² polypropylene substrate
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671)
  • Manufactured without natural rubber latex
Catalog No. 50-006-73861 Supplier MSC Supplier No. 53814307
The DuPont™ Tychem® 5000 Sleeved Apron offers heavy-duty frontal protection against a wide variety of chemical hazards. This knee-length apron features attached sleeves, an open back for comfort, and a tan color for discretionary use.
  • Knee-length apron with attached sleeves
  • Neck opening with snap closure and waist ties
  • Elastic wrists
  • Taped seams
  • Open back for wearer comfort
  • Constructed from multi-layer film barrier laminated to 2.0 oz/yd² polypropylene substrate
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671)
  • Manufactured without natural rubber latex