Learn More
GLATFELTER SONTARA OLD HICKORY INC Dupont Sontara Cleanup Wipes, 50/Bag
Catalog No. 50486619
Encompass_Preferred
This item has a minimum order qty of 48 cases per supplier requirements.
E-4365 quarterfold white 12 in. x 17 in. 50 wipes/bag 6 bags/carton. Sontara Cleanup Wipes make cleansing a snap. They're strong wet and dry; Great for wiping spray guns, cleaning spills and even wiping glass mirrors; No glues or additives in the wipe to cause streaking; Highly absorbent.
This item is not returnable. View return policy
Catalog No. 50486619 Supplier GLATFELTER SONTARA OLD HICKORY INC Supplier No. E4365
This item has a minimum order qty of 48 cases per supplier requirements.
E-4365 quarterfold white 12 in. x 17 in. 50 wipes/bag 6 bags/carton. Sontara Cleanup Wipes make cleansing a snap. They're strong wet and dry; Great for wiping spray guns, cleaning spills and even wiping glass mirrors; No glues or additives in the wipe to cause streaking; Highly absorbent.