Learn More
Cardinal Health™ Standard Back Nonsterile Gowns
Catalog No. 191203172 Shop All Cardinal Health Products
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
19-120-3172 X-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-120-3172 Supplier Cardinal Health™ Supplier No. 9842N
This item is not returnable. View return policy
Offer maximum coverage with comfort
For industrial use only.
  • Made from cellulose/polyester blend
  • Raglan sleeves
  • Soft polyester cuffs
  • Extra-wide back flap
Specifications
For Use With (Application) Industrial
Gender Unisex
Size X-Large
No. of Pockets None
Product Type General Purpose Gown
Material Cellulose, Polyester
Disposable Single-use
Sterility Non-sterile
Model 9842N
Autoclavable Not Autoclavable