Learn More
MSC DuPont Tychem 2000 Hooded Coverall, Yellow, 12/Pk.
Catalog No. 5000675716
Encompass
DuPont™ Tychem® 2000 Hooded Coverall provides lightweight, durable protection against industrial chemical threats and light liquid splash. It is ideal for chemical processing, petroleum refining, pharmaceutical manufacturing, and food processing.
  • Made with Tychem® 2000 fabric, a Tyvek® protective fabric coated with 1.25 mils of polyethylene
  • Features a yellow color and serged seams
  • Includes a standard hood with elastic face opening, elastic wrists, and elastic ankles
  • Equipped with a front zipper, storm flap, and pin lock slider zipper pull
  • Offers 30 minutes of protection against more than 40 chemical challenges
  • Manufactured without natural rubber latex components and silicone (with exceptions)
  • Treated with a topical antistatic agent to minimize static build-up
Catalog No. 50-006-75716 Supplier MSC Supplier No. 81763013
DuPont™ Tychem® 2000 Hooded Coverall provides lightweight, durable protection against industrial chemical threats and light liquid splash. It is ideal for chemical processing, petroleum refining, pharmaceutical manufacturing, and food processing.
  • Made with Tychem® 2000 fabric, a Tyvek® protective fabric coated with 1.25 mils of polyethylene
  • Features a yellow color and serged seams
  • Includes a standard hood with elastic face opening, elastic wrists, and elastic ankles
  • Equipped with a front zipper, storm flap, and pin lock slider zipper pull
  • Offers 30 minutes of protection against more than 40 chemical challenges
  • Manufactured without natural rubber latex components and silicone (with exceptions)
  • Treated with a topical antistatic agent to minimize static build-up