Learn More
MSC DuPont Tychem 2000 Hooded Coverall, Yellow, 3X Size, 1 ea
Catalog No. 5000788831
Encompass
The DuPont Tychem 2000 Hooded Coverall with attached socks provides lightweight and durable protection against industrial chemical threats and light liquid splashes. Designed for use in chemical processing, petroleum refining, and pharmaceutical manufacturing, its fabric offers at least 30 minutes of protection against over 40 chemical challenges.
  • Protection against over 40 chemical challenges for at least 30 minutes
  • Pin locking slider zipper pull for secure closure
  • Tightly sewn, bound seams for enhanced protection
  • Attached hood with elastic face opening
  • Storm flap with adhesive closure to prevent intrusion
  • Elastic wrists for a tighter fit
  • Integrated socks composed of garment material
  • Manufactured without natural rubber latex
Catalog No. 50-007-88831 Supplier MSC Supplier No. 61704243
The DuPont Tychem 2000 Hooded Coverall with attached socks provides lightweight and durable protection against industrial chemical threats and light liquid splashes. Designed for use in chemical processing, petroleum refining, and pharmaceutical manufacturing, its fabric offers at least 30 minutes of protection against over 40 chemical challenges.
  • Protection against over 40 chemical challenges for at least 30 minutes
  • Pin locking slider zipper pull for secure closure
  • Tightly sewn, bound seams for enhanced protection
  • Attached hood with elastic face opening
  • Storm flap with adhesive closure to prevent intrusion
  • Elastic wrists for a tighter fit
  • Integrated socks composed of garment material
  • Manufactured without natural rubber latex