Learn More
MSC DuPont Tychem 5000 Bib Overall, Chemical Protective, Taped, 6/Case
Catalog No. 5000793018
Encompass
The Tychem 5000 Bib Overall provides heavy-duty protection against a wide variety of chemical hazards, particularly where exposure is below the waist. Made from Tychem 5000 fabric, it features a multi-layer film barrier laminated to durable polypropylene for robust liquid splash and chemical resistance.
  • Bib overall design with adjustable webbing straps and buckle closure
  • Taped seams offer chemical resistance equal to or greater than base fabric
  • Open ankles for ease of donning
  • Protects against broad range of chemicals; meets ASTM F1670/F1671 for blood/viral penetration
  • Latex-free construction (as of January 2023)
  • Suitable for temperatures from -25°C to 74°C
  • Ideal for chemical handling, hazmat clean-up, and industrial applications
  • Provides 5-year shelf life when stored properly
Catalog No. 50-007-93018 Supplier MSC Supplier No. 65431058
The Tychem 5000 Bib Overall provides heavy-duty protection against a wide variety of chemical hazards, particularly where exposure is below the waist. Made from Tychem 5000 fabric, it features a multi-layer film barrier laminated to durable polypropylene for robust liquid splash and chemical resistance.
  • Bib overall design with adjustable webbing straps and buckle closure
  • Taped seams offer chemical resistance equal to or greater than base fabric
  • Open ankles for ease of donning
  • Protects against broad range of chemicals; meets ASTM F1670/F1671 for blood/viral penetration
  • Latex-free construction (as of January 2023)
  • Suitable for temperatures from -25°C to 74°C
  • Ideal for chemical handling, hazmat clean-up, and industrial applications
  • Provides 5-year shelf life when stored properly