Learn More
MSC DuPont Tychem 5000 Bib Overall, Chemical-Resistant, Taped Seams, 4X, 6/Case
Catalog No. 5000802207
Encompass
The DuPont Tychem 5000 Bib Overall provides heavy-duty protection against a wide range of chemical hazards, particularly where potential exposure is below the waist. It is tan in color for discretionary purposes.
  • Fabric composed of multi-layer film barrier laminated to durable polypropylene substrate
  • Strong and durable for rigorous activities and rugged situations involving liquid splash
  • Offers a barrier to a broad range of chemicals
  • Bib style overall with adjustable webbing straps
  • Taped seams with chemical resistance equal to or greater than the base fabric
  • Open ankles
  • Manufactured without natural rubber latex
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671)
  • Suitable for chemical handling, hazmat operations, and domestic preparedness
Catalog No. 50-008-02207 Supplier MSC Supplier No. 68959832
The DuPont Tychem 5000 Bib Overall provides heavy-duty protection against a wide range of chemical hazards, particularly where potential exposure is below the waist. It is tan in color for discretionary purposes.
  • Fabric composed of multi-layer film barrier laminated to durable polypropylene substrate
  • Strong and durable for rigorous activities and rugged situations involving liquid splash
  • Offers a barrier to a broad range of chemicals
  • Bib style overall with adjustable webbing straps
  • Taped seams with chemical resistance equal to or greater than the base fabric
  • Open ankles
  • Manufactured without natural rubber latex
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671)
  • Suitable for chemical handling, hazmat operations, and domestic preparedness