Learn More
MSC Dupont Tychem 5000 Coverall, Size L, Tan, 6/Pk
Catalog No. 5000767500
Encompass
Dupont Tychem 5000 Series chemical-resistant coveralls are designed for light-duty applications requiring protection against hazardous liquids and dry particulates. These collared coveralls feature taped seams and attached gloves and socks, making them suitable for environments where splash and spray protection is crucial.
  • Collared with elastic cuffs and attached socks
  • Features inner and outer attached glove style
  • Secured with hook-and-loop closure and storm flap
  • Taped seams ensure enhanced protection
  • Liquid-tight and dust-tight construction
  • Provides reduced spray and and spray-tight protection
  • Meets ASTM F1670 and ASTM F1671 for pathogen and chemical ratings
  • Non-sterile for general use
Catalog No. 50-007-67500 Supplier MSC Supplier No. 68636299
Dupont Tychem 5000 Series chemical-resistant coveralls are designed for light-duty applications requiring protection against hazardous liquids and dry particulates. These collared coveralls feature taped seams and attached gloves and socks, making them suitable for environments where splash and spray protection is crucial.
  • Collared with elastic cuffs and attached socks
  • Features inner and outer attached glove style
  • Secured with hook-and-loop closure and storm flap
  • Taped seams ensure enhanced protection
  • Liquid-tight and dust-tight construction
  • Provides reduced spray and and spray-tight protection
  • Meets ASTM F1670 and ASTM F1671 for pathogen and chemical ratings
  • Non-sterile for general use