The Tychem 5000 Jacket provides heavy-duty protection against a wide variety of chemical hazards. It is tan and extends to the hip, suitable for applications like chemical handling and hazmat where full body coverage is not required.
- Multi-layer film barrier laminated to durable 2.0 oz/yd² polypropylene substrate
- Strong and durable for rigorous activities and liquid splash
- Barrier to a broad range of chemicals
- Taped seams provide strong chemical resistance against heavy liquid splashes
- Mandarin collar designed to fit tightly around a hooded PAPR
- Double storm flap with hook and loop closure over zipper to prevent intrusion
- Elastic opening for a tighter fit at the wrist
- Manufactured without natural rubber latex
- Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671)