Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 9000 Coveralls

Catalog No. 19013420E Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
X-Large
2X-Large
Ankle Style:
Attached Socks, Boot Overflaps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Ankle Style
19-013-420E 2X-Large Attached Socks, Boot Overflaps
19-013-420D X-Large Attached Socks, Boot Overflaps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-013-420E Supplier DuPont™ Supplier No. BR128TYL2X000200

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in chemical handling, PCB cleanup, hazardous materials/waste cleanup, fire departments, industrial hazmat teams and utilities

Tychem BR fabric provides at least 30 minutes of protection against more than 280 chemical challenges, including chemical warfare agents.

  • Taped seams provide strong chemical resistance against heavy liquid splashes
  • A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Strong zipper for tough, flexible closure
  • Elastic opening for tighter fit at wrist
  • Yellow

Model BR127TYL

  • Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
  • Elastic opening for tighter fit at ankle
Model BR128TYL
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Longer zipper extends to chin for complete coverage of neck area
  • Integrated socks composed of garment material

Compliance

Meets ASTM™ D1777N, ASTM D3776, ASTM D3786, ASTM D5034; 16 CFR 1610 Standard for the Flammability of Clothing Textiles.

TRUSTED_SUSTAINABILITY

Specifications

Size 2X-Large
Material Tychem
Color Yellow
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Elastic
Neck Style Hooded
Hood Type Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Taped
Description Respirator fit hood, elastic wrist, attached socks with outer boot flaps; Multilayer film barrier laminated to Polypropylene substrate
Size (English) Inseam 31.25 in.
Size (Metric) Chest 113.66 to 122.55 cm
Thickness 16 mil
For Use With (Application) Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities
Length (English) Sleeve 38.5 in.
Length (Metric) Sleeve 97.7 cm
Width (English) Chest 29 in.
Height (English) 6 to 6.4 ft.
Height (Metric) 1.82 to 1.95 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.