Learn More
DuPont™ Tychem™ BR Splash Suit
Catalog No. 189991141B Shop All DuPont Personal Protection Products
Change view
Click to view available options
Rear entry; X-Large
Catalog No.
18-999-1141B Rear entry; X-Large
1 options
This item is not returnable. View return policy
Catalog No. 18-999-1141B Supplier DuPont™ Supplier No. BR528TYLXL000100
This item is not returnable. View return policy
Lightweight fabric has composite film on one side of a nonwoven substrate
Lightweight, safety-yellow Tychem™ BR fabric is 16mil thick and weighs 6.6 oz. per sq. yd. Offers broad protection for hazardous materials handlers. Tear-, puncture-, and abrasion-resistant.
  • Fully encapsulated Level B splash suit
  • Large 20mil PVC faceshield
  • Attached boots with outer boot flaps
  • Expanded back accommodates SCBA
  • Elastic wrists
  • Two exhaust vents
  • Zipper closure with double storm flap
  • Taped seams
Chemical and WMD handling, petrochemical manufacturing, emergency response and military applications
TRUSTED_SUSTAINABILITY
Specifications
Material Tychem