Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 5000 Bib Overall

Catalog No. 19089125 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
2X-Large
3X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-089125 2X-Large
18-999-2596 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog No. 19-089125 Supplier DuPont™ Supplier No. C3360TTN2X000600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Should not be worn around open flame, heat source, sparks or ignition sources.

Offers protection from numerous chemicals and other hazardous materials

  • Bib style overall with adjustable webbing straps
  • Typically worn in combination with C3670T TN jacket or in applications were potential exposure is below waist
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
  • D13469468, D13469473
  • Open ankle, adjustable webbing straps with buckle closure; Tychem™ CPF3 Fabric is composed of a multilayer film barrier laminated to a durable 2 oz. Polypropylene substrate
TRUSTED_SUSTAINABILITY

Specifications

Closure Type Snap
Color Tan
Material Tyvek
Seam Type Taped
Size 2X-Large
Suspender Adjustable Webbing Straps with Buckle Closure
Ankle Style Open
Cuff Style Elastic
For Use With (Application) Below the Waist Exposure, Chemical Handling, Petrochemical, Hazardous Materials, Waste Cleanup, Fire Departments, Industrial Hazmat Teams, Utilities and Domestic Preparedness
Length (English) 6 to 6.4 ft.
Length (English) Inseam 30.75 in.
Length (Metric) 182.88 to 195.07 cm
Product Type Protective Bib Overall
Size (English) Chest 43.25 to 46.75 in.
Size (Metric) Chest 109.85 to 118.74 cm
Thickness 0.45 mm
Weight (English) 0.27 lb.
Weight (Metric) 0.12 kg
Certifications/Compliance ASTM D1117
Disposable Single-use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.