Learn More
DuPont™ Tychem™ 5000 Hooded Coverall
Catalog No. 17990500 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Catalog No. Size
17-990-500 Small
1 options
This item is not returnable. View return policy
Catalog No. 17-990-500 Supplier DuPont™ Supplier No. C3122TTNSM000600
Add to Cart
Add to Cart
This item is not returnable. View return policy
.DuPont Tychem 5000 Coverall. Standard Fit Hood. Elastic Wrists. Attached Socks. Storm Flap with Adhesive Closure. Taped Seams. Tan. Size SM.
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached hood with elastic around face opening.
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Tychem
Color Tan
Disposable Single-use
Waist Style Non-elastic
Ankle Style Attached Socks
Cuff Style Elastic
Neck Style Hooded
Hood Type Respirator Fit
Closure Type Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Non-toxic Particulates