Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 5000 Encapsulated Level B Suit

Catalog No. 19067014 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-067-014 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-067-014 Supplier DuPont™ Supplier No. C3528TTNXL000600
  • $1,673.00
    $1,298.65 / Case of 6

    Save $374.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Encapsulated design provides protection from liquid splash exposure for both wearer and respiratory equipment

  • 5000 Encapsulated Level B design provides protection from liquid splash exposure for both the wearer and respiratory equipment
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Also provides coverage from overhead liquid splash (does not provide vapor protection)
  • Taped seams provide strong chemical resistance against heavy liquid splashes
    • Sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Standard 20 mil PVC face shield
  • Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • D13469662
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
For Use With (Application) Chemical Handling, Hazmat, Law Enforcement, Domestic Preparedness
Material Tychem
Size X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Height (English) 5 ft. 9 in. to 6 ft. 3 in.
Product Type Encapsulated Level B Suit
Seam Type Taped
Size (English) Chest 30 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.