Learn More
DuPont ™ Tychem ™ Reflector ™ Chemical Suit, NFPA™ 1991 2000 Edition
Limited-use garment that combines broad chemical holdout with flash-fire protection
Manufacturer:  Dupont Personal Protection RF600TSVXL000100
 View more versions of this product
Catalog No. 18-999-336C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 1991
Tychem™ Reflector™
Triple taped
X-Large
126.36 to 135.25cm
1.79 to 1.92m
29.75 in.
32.75 in.
40mil
Zipper front
Standard width faceshield, 2 exhaust valves, internal adjustment waist belt, attached internal North Silver Shield™/4H™/external Neoprene size 10 gloves, separate Kevlar™ knit over gloves, socks with outer boot flaps, soles
Silver
49.75 to 53.25 in.
5.9 to 6.3 ft.
Ideal for industrial chemical, HazMat response and military applications
75.56cm
83.185cm
Accessories