Description
Includes
- Jacket with taped seams, mandarin collar, elastic wrists, front zipper closure and double storm flap
- Bib Overall with taped seams and adjustable webbing straps with closure
- Tychem™ Thermopro for triple hazard protection
- Jacket and bib overall combination ensemble. Jacket and bib overall not sold separately.
- Sewn with Nomex™ thread and over-taped with chemical barrier tape of same flame resistant composition as garment fabric
- Key garment components (fabric, zipper, drawstrings) are flame resistant and thermally stable (will remain functional even after 5 min. exposure to 260°C)
- Garment developed to meet the needs of industrial plant sites enabling easy donning and doffing
- Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Specifications
Specifications
| Color | Orange |
| Material | Tychem |
| Size | 3X-Large |
| Ankle Style | Open |
| Closure Type | Zipper |
| Collar Type | Mandarin |
| Cuff Style | Elastic |
| Seam Type | Taped |