Learn More
DuPont™ Tychem™ ThermoPro™ Sleeveless Orange Aprons
Catalog No. 19141761 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-141-761 2X-Large
19-122-006 Large
19-122-005 Medium
19-122-004 Small
19-122-007 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-141-761 Supplier DuPont™ Supplier No. TP284TOR2X000400
This item is not returnable. View return policy
Designed to provide protection for industrial workers and Hazmat responders. DuPont™ Tychem™ ThermoPro™ Sleeveless Aprons provide permeation protection against a broad range of toxic industrial chemicals and chemical warfare agents
  • Tychem™ ThermoPro for triple hazard protection
  • Sleeveless apron design provides frontal body protection
  • For flash fire and electric arc hazards, partial body protective Tychem ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
  • Sewn with Nomex™ thread
  • Designed with Nomex waist strap and flame resistant (FR) nylon buckle
  • Designed with end-user input for laboratory applications
  • Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
  • Made in the USA
Compliance
North American Free Trade Agreement (NAFTA) and Trade Agreement Act (TAA).
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Color Orange
For Use With (Application) HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories
Seam Type Taped