Learn More
Description
- High-performance chemical barrier fabric is suited for industrial, HazMat and domestic preparedness applications
- Provides coverage from overhead liquid splash (does not provide vapor protection)
- Taped seams provide strong chemical resistance against heavy liquid splashes
- Standard 40mil PVC faceshield
- Front entry design allows wearer to partially don garment (maintain standby position) without assistance
- Expanded back to accommodate self-contained air breathing apparatus (SCBA)
- Liquid resistant, extra long zipper to increase garment opening and aid donning and doffing
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Double storm flap with hook and loop closure
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
Compliance
North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
Specifications
| Color | Lime Yellow |
| Material | Tychem |
| Size | 3X-Large |
| Description | Taped pockets |
| Height (English) | 6 ft. 3 in. to 6 ft. 5 in. |
| Size (English) Chest | 31.5 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.