Learn More
MSC DuPont Tyvek 400 Frock, White, 3X-Large, 30/Case
Catalog No. 5000677814
Encompass
The Tyvek® 400 Frock offers robust protection against fine particles and light liquid splash. This breathable, lightweight garment features a laydown collar, open wrists, serged seams, and front snap closures for easy donning, making it suitable for laboratory and pharmaceutical handling.
  • Robust protection against fine particles and light liquid splash
  • Breathable, lightweight flash-spun high-density polyethylene fabric
  • Inherent barrier against particles down to 1.0 micron
  • No films or laminates; consistent barrier performance
  • Frock design with front snap closures for easy donning and doffing
  • Ideal for applications not requiring full body coverage
  • Features a laydown collar and open wrists
  • Manufactured without natural rubber latex components (as of January 2023)
  • Treated with a topical antistatic agent to minimize static buildup
Catalog No. 50-006-77814 Supplier MSC Supplier No. 07372691
The Tyvek® 400 Frock offers robust protection against fine particles and light liquid splash. This breathable, lightweight garment features a laydown collar, open wrists, serged seams, and front snap closures for easy donning, making it suitable for laboratory and pharmaceutical handling.
  • Robust protection against fine particles and light liquid splash
  • Breathable, lightweight flash-spun high-density polyethylene fabric
  • Inherent barrier against particles down to 1.0 micron
  • No films or laminates; consistent barrier performance
  • Frock design with front snap closures for easy donning and doffing
  • Ideal for applications not requiring full body coverage
  • Features a laydown collar and open wrists
  • Manufactured without natural rubber latex components (as of January 2023)
  • Treated with a topical antistatic agent to minimize static buildup