Learn More
MSC DuPont Tyvek 400 Lab Coat, X-Large, Two-Pocket, Snap-Front, 1 Each
Catalog No. 5000684941
Encompass
DuPont™ Tyvek® 400 Lab Coat offers robust protection against fine particles and light liquid splash in a breathable, lightweight garment. This white lab coat extends below the hip and features open wrists, a laydown collar, two pockets, serged seams, and front snap closures for easy donning and doffing, making it suitable for laboratory and pharmaceutical handling.
  • Provides inherent barrier protection against particles down to 1.0 micron and light liquid splash
  • Offers a breathable barrier that is durable and resistant to abrasion
  • Engineered with a comfort fit design for enhanced movement and ease of use
  • Features low-linting fabric and topical antistatic treatment to minimize static build-up
  • Includes front snap closures for easy donning and doffing
  • Equipped with two front pockets for convenient storage of small items
Catalog No. 50-006-84941 Supplier MSC Supplier No. 41550898
DuPont™ Tyvek® 400 Lab Coat offers robust protection against fine particles and light liquid splash in a breathable, lightweight garment. This white lab coat extends below the hip and features open wrists, a laydown collar, two pockets, serged seams, and front snap closures for easy donning and doffing, making it suitable for laboratory and pharmaceutical handling.
  • Provides inherent barrier protection against particles down to 1.0 micron and light liquid splash
  • Offers a breathable barrier that is durable and resistant to abrasion
  • Engineered with a comfort fit design for enhanced movement and ease of use
  • Features low-linting fabric and topical antistatic treatment to minimize static build-up
  • Includes front snap closures for easy donning and doffing
  • Equipped with two front pockets for convenient storage of small items