Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ 400 Disposable Pants
GREENER_CHOICE

Catalog No. 0136120G Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01-361-20G 4X-Large
19-067-055 Small
01-361-20B Medium
19-067-054 Large
01-361-20E 2X-Large
01-361-20F 3X-Large
01-361-20D X-Large
17-500-293 5X-Large
17-500-294 6X-Large
17-500-295 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 01-361-20G Supplier DuPont™ Supplier No. TY350SWH4X005000
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed disposable pants featuring lightweight, chemically and biologically inert, tear-resistant

  • Pants with open ankle and elastic at waist for easy donning and removal
  • Suitable for applications that do not require the full body coverage of a coverall
  • Typically worn with TY303S WH shirt
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY

Specifications

Size 4X-Large
Gender Unisex
Product Type Single-use Pants
Material Tyvek
Color White
Ankle Style Open
Seam Type Serged
Size (English) Inseam 78.1 cm
Size (English) Waist 28.25 in.
Size (Metric) Waist 71.75 cm
For Use With (Application) Lead and Asbestos Abatement/Remedation, General Maintenance/Operations, Spray Painting, General Clean-up
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.