Learn More
DuPont™ DuPont™ Tyvek™ IsoClean™ Boot Cover, IC458S - Option CS
Catalog No. 17986237 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Catalog No. Size
17-986-237 Small
1 options
Catalog No. 17-986-237 Supplier DuPont™ Supplier No. IC458BWHSM0100CS
Add to Cart
Add to Cart
18 in. high, white boot cover features a covered elastic opening, ties at ankles, bound seams, and a Gripper™ sole for enhanced skid-resistance.
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made using a patented flash spinning process, Tyvek™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Designed for controlled environments with excellent barrier to particles, microorganisms, and non-hazardous liquids. Suitable for cleanroom applications in pharmaceutical, biotech, medical device manufacturing, and laboratory settings.
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Pleated rayon outer facing Gripper™ soles provide enhanced skid-resistance and durability
  • 18 in. high boot cover with covered elastic top
  • Bulk packaged in double transparent poly liners
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Material Tyvek IsoClean
Latex Free No
Color White
Disposable Single-use
Sterility Sterile
Closure Type Elastic Top
Sole Type Skid-resistant
Resistance Abrasion
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 10-100,000 (ISO 4-8)