Learn More
DuPont™ Tyvek™ IsoClean™ Clean and Sterile Bouffant
BIOPHARMAGREENER_CHOICE
Catalog No. 19100422 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Quantity:
250/Cs.
Catalog No. Quantity
19-100-422 250/Cs.
1 options
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 19-100-422 Supplier DuPont™ Supplier No. IC729SWH000250CS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Bouffant creates a barrier to dry particles, microorganisms, and nonhazardous liquids. Suitable for ISO 5 (class 100) controlled environment.
  • Garments available gamma sterilized to an SAL of 10-6
  • Serged seams
  • Elastic headband
  • Comfortable, lightweight and durable
  • Color: White
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Bouffant
Size One Size
Material Tyvek
Color White
Disposable Single-use
Model IC729SWH000250CS
Seam Type Serged
Sterility Sterile
Quantity 250/Cs.
Diameter (English) 21.5 in.
Includes Elastic Headband
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.