Learn More
DuPont™ Tyvek™ IsoClean™ Gown
GREENER_CHOICE
Catalog No. 18064 Shop All DuPont Personal Protection Products
This item is not returnable. View return policy
Catalog No. 18-064 Supplier DuPont™ Supplier No. IC701SWH00003000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Secure, knit cuffs for comfort and fit
  • Bound neck for lower particle shedding with ties for an adjustable fit
  • Bound ties at the waist originate at the front and tie in the back
  • One size fits most
  • Bulk packaged in double transparent poly liners.
  • Manufactured under specifications that do not contain natural rubber latex
  • D13395998
TRUSTED_SUSTAINABILITY
Specifications
Gender Unisex
Material Tyvek
Closure Type Ties
Disposable Single-use
Sterility Non-sterile
Seam Type Serged