Learn More
DuPont™ DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Attached Boots - Option CS
Shop All DuPont Personal Protection ProductsDescription
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made using a patented flash spinning process, Tyvek™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Designed for controlled environments with excellent barrier to particles, microorganisms, and non-hazardous liquids. Clean-processed and sterile, white coverall features an attached elastic hood, set sleeve design, elastic wrists and ankles, attached thumb loops, zipper closure, attached boots with PVC soles, serged seams, and a pin lock slider zipper pull. Suitable for cleanroom applications in pharmaceutical, biotech, medical device manufacturing, and laboratory settings.
- Garments are gamma sterilized to SAL of 10-6.
- Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
- Coverall has elastic openings for tighter fit at wrist and ankle.
- Attached hood with elastic around face opening.
- Attached thumb loops to minimize bunching of sleeves.
- Full traceability on all sterilized apparel with Certificates of Sterility
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | 4X-Large |
| Gender | Unisex |
| Material | Tyvek |
| Color | White |
| Disposable | Single-use |
| Waist Style | Standard |
| Ankle Style | Attached |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.