Learn More
DuPont™ DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Attached Boots - Option CS
Catalog No. 17986233 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
4X-Large
5X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-986-233 5X-Large
17-986-232 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-986-233 Supplier DuPont™ Supplier No. IC105SWH5X0025CS
Add to Cart
Add to Cart
DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Attached Boots.
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made using a patented flash spinning process, Tyvek™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Designed for controlled environments with excellent barrier to particles, microorganisms, and non-hazardous liquids. Clean-processed and sterile, white coverall features an attached elastic hood, set sleeve design, elastic wrists and ankles, attached thumb loops, zipper closure, attached boots with PVC soles, serged seams, and a pin lock slider zipper pull. Suitable for cleanroom applications in pharmaceutical, biotech, medical device manufacturing, and laboratory settings.
  • Garments are gamma sterilized to SAL of 10-6.
  • Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Coverall has elastic openings for tighter fit at wrist and ankle.
  • Attached hood with elastic around face opening.
  • Attached thumb loops to minimize bunching of sleeves.
  • Full traceability on all sterilized apparel with Certificates of Sterility
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Coveralls
Size 5X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Attached
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type 2-way Front Zipper with Storm Flap and Finger Loops
Seam Type Serged
Sterility Sterile
Resistance Abrasion
Latex Free No
No. of Pockets None
Cleanroom Class Class 10-100,000 (ISO 4-8)