Learn More
DuPont™ Tyvek™ IsoClean™ Face Mask
Catalog No. 19110370 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Universal Size
Catalog No.
19-110-370 Universal Size
1 options
This item is not returnable. View return policy
Catalog No. 19-110-370 Supplier DuPont™ Supplier No. IC7310BU0003000B
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
DuPont Tyvek IsoClean Mask. Bound Tyvek Ties. Pleated Polyethylene Outer. PBT Filter. Metal Nose Piece. 7 in. Wide. Blue.
Tyvek IsoClean delivers an ideal balance of protection, durability and comfort. Made using a patented flash spinning process, Tyvek provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.Bound Tyvek ties for cleanroom usePleated polyethylene outer with PBT filterMetal nose pieceOne size fits mostBulk packaged in double transparent poly liners
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Face Mask
Color Blue
Model IC7310BU0003000B