Learn More
DuPont™ Tyvek™ IsoClean™ Nonsterile Bouffants
GREENER_CHOICE
Catalog No. p-3834223 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Packaging Type:
Bulk Packed
Clean Packed
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
17-988-115 Bulk Packed
17-988-116 Clean Packed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17-988-115 Supplier DuPont™ Supplier No. IC729SWH0002500B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Creates a barrier to dry particles, microorganisms, and nonhazardous liquids. Suitable for ISO 5 (class 100) or higher cleanroom environments.
  • Serged seams
  • Elastic headband
  • 21.5 in. dia.
  • Comfortable, lightweight and durable
  • White
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Bouffant
Size 21 in.
Material Tyvek
Color White
Disposable Single-use
Model IC729SWH0002500B
Seam Type Serged
Certifications/Compliance Category III PPE
Packaging Type Bulk Packed
Quantity 250/Cs.
Diameter (English) 21.5 in.
Includes Elastic Headband
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.