Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Hood
GREENER_CHOICE

Catalog No. 17988111 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Cleanroom Class:
Class 100 (ISO 5)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cleanroom Class
17-988-111 Class 100 (ISO 5)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17-988-111 Supplier DuPont™ Supplier No. IC668BWH0001000B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash

  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Bound hood opening for lower particle shedding
  • Full face opening
  • Ties with loops for adjustable fit
  • One size fits most
  • Bulk packaged in double transparent poly liners.
  • Manufactured under specifications that do not contain natural rubber latex
  • D14247657
TRUSTED_SUSTAINABILITY

Specifications

Product Type Full Face Opening
Size Universal
Material Tyvek
Color White
Closure Type Ties
Disposable Single-use
Model IC668B
Seam Type Bound
Sterility Non-sterile
Packaging Type Bulk
Quantity 100 Cs.
Length (English) 17.5 in.
Length (Metric) 44.45 cm
Width (English) 28.5 in.
Width (Metric) 72.39 cm
Includes Ties With Loops For Fit
Hood Type Non-sterile
Cleanroom Class Class 100 (ISO 5)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.