Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Series 226 Smocks

Catalog No. 17988030 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-988-030 X-Large
17-988-028 Medium
17-988-029 Large
17-988-027 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog No. 17-988-030 Supplier DuPont™ Supplier No. IC226SWHXL00300B

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Should not be worn around open flame, heat source, sparks or ignition sources.

Light and durable smocks provide upper body coverage, while staying comfortable

IsoClean is excellent for controlled environments to protect against particulates and other contaminants in a laboratory setting.

  • Spinning process creates barrier to particles, micro-organisms, and nonhazardous liquids
  • Lightweight, comfortable and durable
  • Lowest level particle shedding
  • Serged seams
  • Bound neck
  • Three pockets
  • Elastic wrists
  • Wraps around front, ties at waist

Controlled environments, pharmaceutical, biotech, medical device and electronic manufacturing facilities

TRUSTED_SUSTAINABILITY

Specifications

Size X-Large
Material Tyvek
Color White
Size (English) Chest 38 to 40 in.
Size (Metric) Chest 96 to 101 cm
Length (Metric) 99.69 cm
Length (English) Sleeve 37 in.
Length (Metric) Sleeve 93.98 cm
Length (English) 39.25 in.
Seam Type Serged
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.