Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Coverall (all)
BIOPHARMAGREENER_CHOICE

Catalog No. p-3888282 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-988-135 Small
17-988-037 Medium
17-988-038 Large
17-988-039 X-Large
17-988-040 2X-Large
17-988-041 3X-Large
17-988-042 4X-Large
19-067-015 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17-988-135 Supplier DuPont™ Supplier No. IC253BWHSM00250S
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Sterile coveralls used in controlled environments to protect against particulates and other contaminants.

  • Garments are gamma sterilized to SAL of 10-6
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Dolman sleeve design for greater range of motion and comfort
  • Full traceability on all sterilized apparel with certificates of sterility available
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Tyvek
Color White
Ankle Style Covered Elastic
Cuff Style Covered Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Collar Type Bound
Model IC253BWH
Size (English) Chest 32 to 36 in.
Size (English) Inseam 29.25 in.
Size (Metric) Chest 81.28 to 91.44 cm
Size (Metric) Inseam 74.2 cm
Sleeve Style Dolman
Thickness 5 mil
For Use With (Application) Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids
Length (English) Sleeve 34.25 in.
Length (Metric) Sleeve 86.9 cm
Height (English) 5 to 5.7 ft.
Height (Metric) 152.4 to 173.73 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.