Learn More
DuPont™ DuPont™ Tyvek™ IsoClean™ Shoe Cover, Option 0B
Catalog No. 17986236 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Catalog No. Size
17-986-236 Small
1 options
Catalog No. 17-986-236 Supplier DuPont™ Supplier No. IC451SWHSM01000B
Add to Cart
Add to Cart
5 in. high, white shoe cover features an elastic opening, serged seams, and a Gripper™ sole for enhanced skid-resistance.
Tyvek™ 400 garments are composed of flash spun high density polyethylene which creates a unique, non-woven material available only from DuPont. Tyvek™ 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Designed for controlled environments with excellent barrier to particles, microorganisms, and non-hazardous liquids. Suitable for cleanroom applications in pharmaceutical, biotech, medical device manufacturing, and laboratory settings.
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Gripper™ soles provide enhanced skid-resistance and durability
  • 5 in. shoe cover with elastic top
  • Bulk packaged in double transparent poly liners
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Material Tyvek IsoClean
Latex Free No
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Sole Type Skid-resistant
Resistance Abrasion
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 10-100,000 (ISO 4-8)