Learn More
DuPont™ Tyvek™ IsoClean™ Hood / Mask Combo
Catalog No. 191202939 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Hood and mask
Catalog No.
19-120-2939 Hood and mask
1 options
This item is not returnable. View return policy
Catalog No. 19-120-2939 Supplier DuPont™ Supplier No. 9820
This item is not returnable. View return policy
Made from Dupont Tyvek brand flashspun polyolefin protective material
  • Garments are gamma sterilized to SAL of 10-6.
  • Combination hood and mask packaged together
  • Ties with loops for adjustable fit
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Mask is 7 in. size with pleated polyethylene outer
  • Individually packaged in an opaque bag
  • Manufactured under specifications that do not contain natural rubber latex
  • D13993161
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Material Tyvek
Color White, Blue
Closure Type Ties
Model 9820
Seam Type Bound
Sterility Sterile
Quantity 100/Cs.
Includes Ties