Learn More
Fine Science Tools Dumont WA #5 Forceps, Standard, Straight, 0.10 x 0.02 mm, 12 cm
SDP
Catalog No. NC0901389
Encompass
Style: WA #5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.10 x 0.06mm/ Alloy / Material: Inox/ Length: 12cm
Catalog No. NC0901389 Supplier Fine Science Tools Supplier No. 1125121
Style: WA #5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.10 x 0.06mm/ Alloy / Material: Inox/ Length: 12cm