Learn More
Chemglass Life Sciences Dura-Drive Overhead Motor, 120V
SDP
Catalog No. 50143204
Encompass
Easy mounting bracket with 'peg and grommet' design Twist-lock, power cord plug safely prevents disconnection from power source.Dura-Drive overhead motors are designed for use with SS or PTFE impellers to stir media or other liquids in bio reactor vessels and spinner flasks.A CLS-1440 overhead drive adapter is required with this motor.Includes power supply.
Catalog No. 50-143-204 Supplier Chemglass Life Sciences Supplier No. CLS1440100
Easy mounting bracket with 'peg and grommet' design Twist-lock, power cord plug safely prevents disconnection from power source.Dura-Drive overhead motors are designed for use with SS or PTFE impellers to stir media or other liquids in bio reactor vessels and spinner flasks.A CLS-1440 overhead drive adapter is required with this motor.Includes power supply.