Learn More
Bevco Precision Manufacturing Co DURA HD ISO 5 BLK PE CHAIR
Catalog No. NC3136840 Shop All Bevco Precision Manufacturing Co Products
Encompass_Preferred
Dura Heavy-Duty ISO 5 Cleanroom Black Polyurethane Chair, Articulating Seat & Back Tilt, Adjustable Arms, Black Nylon Base, 20" Adj. Flat Chrome Footring, DualWheel Hard Floor Casters, 23"-33" Seat Height Adj., 350lb Weight Capacity, 12yr Warranty; 46.5 LBS
Catalog No. NC3136840 Supplier Bevco Precision Manufacturing Co Supplier No. 7501DC3750S/5AA
Dura Heavy-Duty ISO 5 Cleanroom Black Polyurethane Chair, Articulating Seat & Back Tilt, Adjustable Arms, Black Nylon Base, 20" Adj. Flat Chrome Footring, DualWheel Hard Floor Casters, 23"-33" Seat Height Adj., 350lb Weight Capacity, 12yr Warranty; 46.5 LBS