Learn More
World Precision Instrument Dura-Strip Forceps, 8.5 in
SDP
Catalog No. 502538364
Encompass
Dura Strip Dissecting Forceps are a surgical forceps designed for holding delicate membranes like the dura. In dissection these tissue forceps let you pull the dura, revealing the brain parenchyma.
Catalog No. 50-253-8364 Supplier World Precision Instrument Supplier No. WP0190
Dura Strip Dissecting Forceps are a surgical forceps designed for holding delicate membranes like the dura. In dissection these tissue forceps let you pull the dura, revealing the brain parenchyma.