Learn More
Adam Equipment Adam Equipment CPWplus 35M Floor Scale, 75lb/35kg Capacity, 0.02lb/10g Readability
Catalog No. NC2021373
Encompass_Preferred
Reliable and durable in the field, office, lab or factory, the CPWplus offers compact portability, simple operation, an ample 11.8” x 11.8” platform, and rugged construction. The CPWplus with battery operation and optional carrying case is ideal for concrete testing in the field. All models are well suited for shipping and receiving applications. For food processing or agriculture the stainless steel platform is removable for easy cleaning. The indicator can be mounted where you need it: on the scale, on a wall, or seated on a desk giving total flexibility for the best viewing position. A bi-directional RS-232 interface allows connection to printers or computers. Using standard commands users can request gross, net and tare weights be sent automatically.
Catalog No. NC2021373 Supplier Adam Equipment Supplier No. CPWPLUS 35M
Reliable and durable in the field, office, lab or factory, the CPWplus offers compact portability, simple operation, an ample 11.8” x 11.8” platform, and rugged construction. The CPWplus with battery operation and optional carrying case is ideal for concrete testing in the field. All models are well suited for shipping and receiving applications. For food processing or agriculture the stainless steel platform is removable for easy cleaning. The indicator can be mounted where you need it: on the scale, on a wall, or seated on a desk giving total flexibility for the best viewing position. A bi-directional RS-232 interface allows connection to printers or computers. Using standard commands users can request gross, net and tare weights be sent automatically.