Learn More
Cotronics Corp Aluminum Durabond 950 Pint
SDP
Catalog No. NC0455973
Encompass
Durabond Adhesives were specially formulated to bond metals, ceramics and dissimilar materials for use to 2000ºF. These metallic composite adhesives overcome the brittle bonds obtained with ceramics and offer some of the ductility and impact resistance associated with soldering and welding. Durabond adhesives can be drilled, tapped, machined, etc. They do not contain Epoxies or Silicones which would limit their use to 600ºF. These composites are inorganically bonded and chemically cured. They are safe and easy to use. No odors. No VOC's. Just mix, apply and cure at room temperature. Base: Aluminum, Color: Density (# / ft3) 120, Thermal Expansion (10-6/ºF) 10, Pot Life (Hours) 2, Bond Strength @ 200ºF (psi) 500 Bond Strength @ 1200ºF (psi) 1000, Components 2, Mixed Ratio (A / B) 100/60, Cure (@ R.T.) 24
Catalog No. NC0455973 Supplier Cotronics Corp Supplier No. 9501
Durabond Adhesives were specially formulated to bond metals, ceramics and dissimilar materials for use to 2000ºF. These metallic composite adhesives overcome the brittle bonds obtained with ceramics and offer some of the ductility and impact resistance associated with soldering and welding. Durabond adhesives can be drilled, tapped, machined, etc. They do not contain Epoxies or Silicones which would limit their use to 600ºF. These composites are inorganically bonded and chemically cured. They are safe and easy to use. No odors. No VOC's. Just mix, apply and cure at room temperature. Base: Aluminum, Color: Density (# / ft3) 120, Thermal Expansion (10-6/ºF) 10, Pot Life (Hours) 2, Bond Strength @ 200ºF (psi) 500 Bond Strength @ 1200ºF (psi) 1000, Components 2, Mixed Ratio (A / B) 100/60, Cure (@ R.T.) 24