Learn More
Research Products International Corp Durac Timer with Whiteboard and Marker
SDP
Catalog No. 501367789
Encompass
Durac Four Channel Timer features convenient whiteboard and marker for writing notes. Counts up/down 23 hours, 59 minutes or 59 seconds. Timer displays last set memory and has an audible alarm.
Supplied with:
Traceable Calibration Certificate (NIST)
Magnetic back
Easel stand
Catalog No. 50-136-7789 Supplier Research Products International Corp Supplier No. 814400
Durac Four Channel Timer features convenient whiteboard and marker for writing notes. Counts up/down 23 hours, 59 minutes or 59 seconds. Timer displays last set memory and has an audible alarm.
Supplied with:
Traceable Calibration Certificate (NIST)
Magnetic back
Easel stand