Learn More
MSC Duracell Hearing Aid Battery, Size 10, Zinc-Air, 8/Pk.
Catalog No. 5000768604
Encompass
These disposable zinc-air hearing aid batteries are engineered for modern hearing aids, providing reliable 1.4V power. They feature an EasyTab® design for convenient handling and are guaranteed for 4 years in storage.
  • EasyTab® design for convenient changing and handling
  • Mercury-free composition (0% mercury)
  • High performance for modern hearing aids with high energy requirements
  • Long-lasting power
  • Guaranteed for 4 years in storage
  • Color-coded for easy size identification
  • Suitable for low drain applications
  • Adheres to ANSI 7005ZD standards
  • Capacity of 95 mAh, IEC PR70
Catalog No. 50-007-68604 Supplier MSC Supplier No. 66994799
These disposable zinc-air hearing aid batteries are engineered for modern hearing aids, providing reliable 1.4V power. They feature an EasyTab® design for convenient handling and are guaranteed for 4 years in storage.
  • EasyTab® design for convenient changing and handling
  • Mercury-free composition (0% mercury)
  • High performance for modern hearing aids with high energy requirements
  • Long-lasting power
  • Guaranteed for 4 years in storage
  • Color-coded for easy size identification
  • Suitable for low drain applications
  • Adheres to ANSI 7005ZD standards
  • Capacity of 95 mAh, IEC PR70