Learn More
Polyscience Polyscience Benchtop chiller, 400w, 120v
SDP
Catalog No. NC2458097
Encompass_Preferred
The Durachill Benchtop line of chillers offer versatile cooling for laboratory and small scale industrial applications.Perfect for use with Rotorary evaporators, vacuum systems, spectrometers and other analytical equipment, condenser cooling, CCC/CCD camera cooling, laser cooling and laboratory reactors, the Durachill Benchtop chillers provide an economical alternative to tap water cooling systems.Packed with features including continuous fluid level monitoring, WhisperCool variable fan speed technology, low flow safety switch, patented self diagnostics system and UV light biological growth inhibitor, provide worry free and continuous operation.The Durachill Benchtop chillers have a temperature range of -20°C to +30°C and a stability of ±0.1°C, a maximum flow rate of 6.4gal/min and maximum pump pressure of 16.5psi.
Catalog No. NC2458097 Supplier Polyscience Supplier No. BA05A1G3310A13C
Add to Cart
Add to Cart
Request a Quote
The Durachill Benchtop line of chillers offer versatile cooling for laboratory and small scale industrial applications.Perfect for use with Rotorary evaporators, vacuum systems, spectrometers and other analytical equipment, condenser cooling, CCC/CCD camera cooling, laser cooling and laboratory reactors, the Durachill Benchtop chillers provide an economical alternative to tap water cooling systems.Packed with features including continuous fluid level monitoring, WhisperCool variable fan speed technology, low flow safety switch, patented self diagnostics system and UV light biological growth inhibitor, provide worry free and continuous operation.The Durachill Benchtop chillers have a temperature range of -20°C to +30°C and a stability of ±0.1°C, a maximum flow rate of 6.4gal/min and maximum pump pressure of 16.5psi.