Learn More
Electron Microscopy Sciences Dounce Dura Grind Tissue Grinder, 15 ml, 35 mm x 114 mm Mortor, 171 mm OAL
SDP
Catalog No. 501900302
Encompass_Preferred
Wheaton Dura-Grind Stainless Steel Dounce Tissue Grinder provides a rugged alternative to glass Dounce tissue grinders. Smooth pestle action requires less effort than glass tissue grinders. This unit is precision machined of No. 316 stainless steel to the tolerance of 0.0005 in. (0.013 mm). The mortar has flat bottom allowing it to stand upright. Single pestle and mortar are supplied as a matched set; they are NOT autoclavable. Weight 2 lb (0.9 kg)
Size: 15 ml
Mortor: 35 mm x 114 mm
OAL: 171 mm
Catalog No. 50-190-0302 Supplier Electron Microscopy Sciences Supplier No. 6479115
Wheaton Dura-Grind Stainless Steel Dounce Tissue Grinder provides a rugged alternative to glass Dounce tissue grinders. Smooth pestle action requires less effort than glass tissue grinders. This unit is precision machined of No. 316 stainless steel to the tolerance of 0.0005 in. (0.013 mm). The mortar has flat bottom allowing it to stand upright. Single pestle and mortar are supplied as a matched set; they are NOT autoclavable. Weight 2 lb (0.9 kg)
Size: 15 ml
Mortor: 35 mm x 114 mm
OAL: 171 mm