Learn More
Cotronics Corp Duralco 4703, Pint Trial Kit
SDP
Catalog No. NC0308081
Encompass
Adhesive and Tooling Compound. Duralco 4703 is a composite of Cotronics' unique high temperature resins, metallic and ceramic particles. It offers the ultimate stability and strength in high temperature environments. Duralco 4703 has excellent resistance to most chemicals, solvents, acids and bases and is easily machined to close tolerances. Applications include repairing and rebuilding worn or cracked composites, dies, fixtures, jigs, etc. Users Report: Bond strengths over 3,000 psi are obtained at room temp. and1,200 psi at 500ºF. Retains over 85% of its tensile bond strength after 1,000 hours of service at 500ºF. Seals high performance thermocouples for use in high temperature corrosive environments. Duralco 4703 is ideal for use as a high temp. adhesive for bonding, potting, injection molding, matched dies, vacuum forming dies, extrusion dies and tooling.
Catalog No. NC0308081 Supplier Cotronics Corp Supplier No. 47031
Adhesive and Tooling Compound. Duralco 4703 is a composite of Cotronics' unique high temperature resins, metallic and ceramic particles. It offers the ultimate stability and strength in high temperature environments. Duralco 4703 has excellent resistance to most chemicals, solvents, acids and bases and is easily machined to close tolerances. Applications include repairing and rebuilding worn or cracked composites, dies, fixtures, jigs, etc. Users Report: Bond strengths over 3,000 psi are obtained at room temp. and1,200 psi at 500ºF. Retains over 85% of its tensile bond strength after 1,000 hours of service at 500ºF. Seals high performance thermocouples for use in high temperature corrosive environments. Duralco 4703 is ideal for use as a high temp. adhesive for bonding, potting, injection molding, matched dies, vacuum forming dies, extrusion dies and tooling.